Product Details

SNP ID
rs1106226
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:1535512 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GCAGCCAGGTCCACGAGCCATCTGC[G/A]GGGGGACAGGGGCGGTCAGGCGGCT
Phenotype
MIM: 612316 MIM: 616101
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ATAD3A PubMed Links

Gene Details

Gene
ATAD3A
Gene Name
ATPase family, AAA domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170535.1 Intron NP_001164006.1
NM_001170536.1 Intron NP_001164007.1
NM_018188.3 Intron NP_060658.3
XM_017001628.1 Intron XP_016857117.1
Gene
SSU72
Gene Name
SSU72 homolog, RNA polymerase II CTD phosphatase
There are no transcripts associated with this gene.

Gene
TMEM240
Gene Name
transmembrane protein 240
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114748.1 Intron NP_001108220.1

View Full Product Details