Product Details
- SNP ID
-
rs1519659
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:100471630 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATACTCACCTCCCTATTTTTTCCT[A/G]TTTGACTTTATGTTCCCTGAGGGTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NMS
PubMed Links
Gene Details
- Gene
- NMS
- Gene Name
- neuromedin S
View Full Product Details