Product Details

SNP ID
rs1559711
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:104766767 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAATGAGAAAACATGCAGTCTCAT[A/C]TATCAAACCATAAAATTGTGAATTA
Phenotype
MIM: 608871 MIM: 608872
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NIPSNAP3A PubMed Links
Additional Information
For this assay, SNP(s) [rs35350211] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NIPSNAP3A
Gene Name
nipsnap homolog 3A
There are no transcripts associated with this gene.

Gene
NIPSNAP3B
Gene Name
nipsnap homolog 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018376.3 Intron NP_060846.2
XM_011518839.1 Intron XP_011517141.1
XM_017014889.1 Intron XP_016870378.1

View Full Product Details