Product Details

SNP ID
rs1158144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:63654134 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTTTATCGAGTTACCTGCCTAG[C/T]AACTGTCTTCTTTTCTAGACTTAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNTN PubMed Links

Gene Details

Gene
SNTN
Gene Name
sentan, cilia apical structure protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080537.1 Intron NP_001074006.1

View Full Product Details