Product Details

SNP ID
rs843331
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:184243392 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTATTTAGAATTAAGAATCAAATAT[A/C]CTTCAATATGTGAAGTGCTCAGCAC
Phenotype
MIM: 608750 MIM: 610145 MIM: 611620
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ALG3 PubMed Links
Additional Information
For this assay, SNP(s) [rs6806627] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ALG3
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006941.2 Intron NP_001006942.1
NM_005787.5 Intron NP_005778.1
XM_011512322.1 Intron XP_011510624.1
XM_011512323.2 Intron XP_011510625.1
Gene
ECE2
Gene Name
endothelin converting enzyme 2
There are no transcripts associated with this gene.

Gene
MIR1224
Gene Name
microRNA 1224
There are no transcripts associated with this gene.

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
There are no transcripts associated with this gene.

View Full Product Details