Product Details

SNP ID
rs906493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121884466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCTTCATAATTTATTTTGCATTA[C/T]ACATATTCTTAAGCATGTAACAAAT
Phenotype
MIM: 602345
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPC3 PubMed Links

Gene Details

Gene
TRPC3
Gene Name
transient receptor potential cation channel subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130698.1 Intron NP_001124170.1
NM_003305.2 Intron NP_003296.1
XM_011532217.2 Intron XP_011530519.1
XM_011532218.2 Intron XP_011530520.1
XM_017008578.1 Intron XP_016864067.1
XM_017008579.1 Intron XP_016864068.1

View Full Product Details