Product Details

SNP ID
rs1525916
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:109468327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGCTTTGGAGACAAGAATTAGC[A/G]CTGTGACACCAGTTCTTAAATCATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYPL2 PubMed Links

Gene Details

Gene
SYPL2
Gene Name
synaptophysin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040709.1 Intron NP_001035799.1
XM_011541283.2 Intron XP_011539585.1
XM_011541284.2 Intron XP_011539586.1
XM_011541285.1 Intron XP_011539587.1

View Full Product Details