Product Details

SNP ID
rs1059297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33299751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGATTAGACCCGTTACCCCGTGG[C/G]TCTGGCCGACCGTCCTGACTCGGAG
Phenotype
MIM: 605660 MIM: 602306 MIM: 601962
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6834 PubMed Links

Gene Details

Gene
MIR6834
Gene Name
microRNA 6834
There are no transcripts associated with this gene.

Gene
PFDN6
Gene Name
prefoldin subunit 6
There are no transcripts associated with this gene.

Gene
RGL2
Gene Name
ral guanine nucleotide dissociation stimulator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243738.1 3534 Intron NP_001230667.1
NM_004761.4 3534 Intron NP_004752.1
XM_011514781.1 3534 Intron XP_011513083.1
XM_011514785.2 3534 Intron XP_011513087.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003190.4 3534 UTR 3 NP_003181.3
NM_172208.2 3534 Intron NP_757345.2
NM_172209.2 3534 UTR 3 NP_757346.2
XM_011514828.2 3534 UTR 3 XP_011513130.1
XM_017011227.1 3534 UTR 3 XP_016866716.1

View Full Product Details