Product Details
- SNP ID
-
rs10067
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
58
- Location
-
Chr.1:111414689 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GAAGACCCAAGTTACCCATCCTGGG[C/G]TGAGTGCCCACCTCAGAAGTCAAAT
- Phenotype
-
MIM: 603578
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OVGP1
PubMed Links
Gene Details
- Gene
- OVGP1
- Gene Name
- oviductal glycoprotein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002557.3 |
1868 |
Missense Mutation |
CAC,CAG |
H604Q |
NP_002548.3 |
View Full Product Details