Product Details

SNP ID
rs878320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15515272 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAATTCTTTGAGGCAACTTCAGT[C/G]CTCATCCGTAGTCAGCCACACAAAC
Phenotype
MIM: 604063
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITGA8 PubMed Links

Gene Details

Gene
ITGA8
Gene Name
integrin subunit alpha 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291494.1 5597 UTR 3 NP_001278423.1
NM_003638.2 5597 UTR 3 NP_003629.2
XM_011519752.2 5597 Intron XP_011518054.1

View Full Product Details