Product Details

SNP ID
rs1045462
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:11674572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGCGGAACCCCAGGAGGCTGA[A/G]AAGTCGAGGTTGCGGCATCCCTCAC
Phenotype
MIM: 605647 MIM: 604094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO6 PubMed Links

Gene Details

Gene
FBXO6
Gene Name
F-box protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018438.5 888 Intron NP_060908.1
XM_005263448.4 888 Intron XP_005263505.1
XM_005263449.4 888 Intron XP_005263506.1
XM_005263451.4 888 Intron XP_005263508.1
XM_006710571.3 888 Intron XP_006710634.1
XM_011541232.2 888 Intron XP_011539534.1
XM_017001007.1 888 Intron XP_016856496.1
Gene
MAD2L2
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127325.1 888 UTR 3 NP_001120797.1
NM_006341.3 888 UTR 3 NP_006332.3
XM_011540507.1 888 UTR 3 XP_011538809.1

View Full Product Details