Product Details

SNP ID
rs1055730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47164476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCAAAGGTGGGGGCTGGGCTGA[A/G]ACTGCCGGTGCGGCAGGGGAAGATG
Phenotype
MIM: 606908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFGAP2 PubMed Links

Gene Details

Gene
ARFGAP2
Gene Name
ADP ribosylation factor GTPase activating protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242832.1 3297 UTR 3 NP_001229761.1
NM_032389.4 3297 UTR 3 NP_115765.2
XM_005253166.1 3297 UTR 3 XP_005253223.1
XM_005253167.1 3297 UTR 3 XP_005253224.1
XM_005253168.2 3297 UTR 3 XP_005253225.1
XM_006718346.1 3297 UTR 3 XP_006718409.1
XM_011520405.1 3297 UTR 3 XP_011518707.1
XM_017018413.1 3297 UTR 3 XP_016873902.1
XM_017018414.1 3297 UTR 3 XP_016873903.1
XM_017018415.1 3297 UTR 3 XP_016873904.1
Gene
C11orf49
Gene Name
chromosome 11 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003676.2 3297 Intron NP_001003676.1
NM_001003677.2 3297 Intron NP_001003677.1
NM_001003678.2 3297 Intron NP_001003678.1
NM_001278222.1 3297 Intron NP_001265151.1
NM_024113.4 3297 Intron NP_077018.1
XM_006718315.1 3297 Intron XP_006718378.1
XM_011520364.2 3297 Intron XP_011518666.1
XM_011520365.2 3297 Intron XP_011518667.1
XM_017018276.1 3297 Intron XP_016873765.1
XM_017018277.1 3297 Intron XP_016873766.1
XM_017018278.1 3297 Intron XP_016873767.1
XM_017018279.1 3297 Intron XP_016873768.1
XM_017018280.1 3297 Intron XP_016873769.1
XM_017018281.1 3297 Intron XP_016873770.1
XM_017018282.1 3297 Intron XP_016873771.1

View Full Product Details