Product Details
- SNP ID
-
rs1224442
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:45016292 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACGGAGGAATGCTTCCATTCAGAC[A/G]GCCCCAGTAAGTACACCCTTGCTTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DBX2
PubMed Links
Gene Details
- Gene
- DBX2
- Gene Name
- developing brain homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004329.2 |
1186 |
Silent Mutation |
GCC,GCT |
A338A |
NP_001004329.2 |
View Full Product Details