Product Details

SNP ID
rs1268086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125792578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATCTAATTATCATGTCTTTTATC[A/G]CAGGCAAAAGTAGAAATAAAACATG
Phenotype
MIM: 609752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCOA7 PubMed Links

Gene Details

Gene
NCOA7
Gene Name
nuclear receptor coactivator 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122842.2 Intron NP_001116314.1
NM_001199619.1 Intron NP_001186548.1
NM_001199620.1 Intron NP_001186549.1
NM_001199621.1 Intron NP_001186550.1
NM_001199622.1 Intron NP_001186551.1
NM_181782.4 Intron NP_861447.3
XM_005266822.4 Intron XP_005266879.1
XM_006715340.3 Intron XP_006715403.1
XM_011535455.2 Intron XP_011533757.1
XM_017010269.1 Intron XP_016865758.1
XM_017010270.1 Intron XP_016865759.1
XM_017010271.1 Intron XP_016865760.1
XM_017010272.1 Intron XP_016865761.1
XM_017010273.1 Intron XP_016865762.1
XM_017010274.1 Intron XP_016865763.1
Gene
NCOA7-AS1
Gene Name
NCOA7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details