Product Details
- SNP ID
-
rs1051419
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:10639738 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TAGACATTTTGGAGTTCTTGTTCTC[A/G]TAATCCTTGATGGGGACCGTGTTGG
- Phenotype
-
MIM: 601920
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
JAG1
PubMed Links
Gene Details
- Gene
- JAG1
- Gene Name
- jagged 1
- Gene
- MIR6870
- Gene Name
- microRNA 6870
There are no transcripts associated with this gene.
View Full Product Details