Product Details

SNP ID
rs137074
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:42577696 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGGAAGGAATCGCAGGCTTTGTCA[C/G]CCGGACAGTCTCCTACACCACACAC
Phenotype
MIM: 611520
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLDIP3 PubMed Links

Gene Details

Gene
POLDIP3
Gene Name
DNA polymerase delta interacting protein 3
There are no transcripts associated with this gene.

Gene
RRP7BP
Gene Name
ribosomal RNA processing 7 homolog B, pseudogene
There are no transcripts associated with this gene.

Gene
SERHL2
Gene Name
serine hydrolase-like 2
There are no transcripts associated with this gene.

View Full Product Details