Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001136219.1 | Intron | NP_001129691.1 | |||
NM_021642.3 | Intron | NP_067674.2 | |||
XM_011509287.2 | Intron | XP_011507589.1 | |||
XM_011509290.2 | Intron | XP_011507592.1 | |||
XM_011509291.1 | Intron | XP_011507593.1 | |||
XM_017000663.1 | Intron | XP_016856152.1 | |||
XM_017000664.1 | Intron | XP_016856153.1 | |||
XM_017000665.1 | Intron | XP_016856154.1 | |||
XM_017000666.1 | Intron | XP_016856155.1 | |||
XM_017000667.1 | Intron | XP_016856156.1 | |||
XM_017000668.1 | Intron | XP_016856157.1 |