Product Details

SNP ID
rs7984554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48240672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTAAAAATGTTATTTATTCATT[A/G]CATAATAGGTGTTAATAGAATTTGA
Phenotype
MIM: 603904
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITM2B PubMed Links

Gene Details

Gene
ITM2B
Gene Name
integral membrane protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021999.4 Intron NP_068839.1

View Full Product Details