Product Details
- SNP ID
-
rs11987209
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143612883 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTGGGCTCTGCCAGCCGATGGTCC[A/G]CTGGCACCTGATCCTGTCTTCCAGC
- Phenotype
-
MIM: 616408
MIM: 137020
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PYCRL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75900260] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PYCRL
- Gene Name
- pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.
- Gene
- TSTA3
- Gene Name
- tissue specific transplantation antigen P35B
View Full Product Details