Product Details

SNP ID
rs6745809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:109561017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACCTCCTTCACATTAGAGCAAA[G/T]AATAATTTTTAAAATGCAAATATTA
Phenotype
MIM: 601181 MIM: 611737
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RANBP2 PubMed Links

Gene Details

Gene
RANBP2
Gene Name
RAN binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006267.4 Intron NP_006258.3
XM_005264002.2 Intron XP_005264059.1
XM_005264003.2 Intron XP_005264060.1
XM_005264004.2 Intron XP_005264061.1
XM_005264005.4 Intron XP_005264062.1
XM_005264007.2 Intron XP_005264064.1
XM_011511575.2 Intron XP_011509877.1
XM_011511576.2 Intron XP_011509878.1
XM_011511578.2 Intron XP_011509880.1
XM_017004623.1 Intron XP_016860112.1
XM_017004624.1 Intron XP_016860113.1
XM_017004625.1 Intron XP_016860114.1
Gene
SEPT10
Gene Name
septin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321496.1 Intron NP_001308425.1
NM_001321498.1 Intron NP_001308427.1
NM_001321499.1 Intron NP_001308428.1
NM_001321500.1 Intron NP_001308429.1
NM_001321501.1 Intron NP_001308430.1
NM_001321502.1 Intron NP_001308431.1
NM_001321503.1 Intron NP_001308432.1
NM_001321504.1 Intron NP_001308433.1
NM_001321505.1 Intron NP_001308434.1
NM_001321506.1 Intron NP_001308435.1
NM_001321507.1 Intron NP_001308436.1
NM_001321508.1 Intron NP_001308437.1
NM_001321509.1 Intron NP_001308438.1
NM_001321510.1 Intron NP_001308439.1
NM_001321511.1 Intron NP_001308440.1
NM_001321512.1 Intron NP_001308441.1
NM_001321513.1 Intron NP_001308442.1
NM_001321514.1 Intron NP_001308443.1
NM_001321515.1 Intron NP_001308444.1
NM_144710.4 Intron NP_653311.1
NM_178584.3 Intron NP_848699.1
XM_006712317.1 Intron XP_006712380.1
XM_011510698.2 Intron XP_011509000.1
XM_011510699.2 Intron XP_011509001.1
XM_011510700.2 Intron XP_011509002.1
XM_011510701.2 Intron XP_011509003.1
XM_011510702.2 Intron XP_011509004.1
XM_011510703.2 Intron XP_011509005.1
XM_011510704.2 Intron XP_011509006.1

View Full Product Details