Product Details
- SNP ID
-
rs1052961
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:26920031 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCATCTTTCATTATAAAGAGATTA[G/T]TAATATTCACCAATCATGCCAATGA
- Phenotype
-
MIM: 605007
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAMTS5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143398098] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAMTS5
- Gene Name
- ADAM metallopeptidase with thrombospondin type 1 motif 5
View Full Product Details