Product Details
- SNP ID
-
rs1054768
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:111293695 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAATATGTCTTATGGGAAGAGAC[G/T]GACGTAGCACAAATCATTAAAAAAC
- Phenotype
-
MIM: 615693
MIM: 615694
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C11orf53
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115778629] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C11orf53
- Gene Name
- chromosome 11 open reading frame 53
There are no transcripts associated with this gene.
- Gene
- COLCA1
- Gene Name
- colorectal cancer associated 1
- Gene
- COLCA2
- Gene Name
- colorectal cancer associated 2
There are no transcripts associated with this gene.
View Full Product Details