Product Details

SNP ID
rs12782196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15045395 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTATCCCAGCTCTGACTGCCTA[C/T]TTTTCTTGGAAGAACTTGCTCTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105376432 PubMed Links

Gene Details

Gene
LOC105376432
Gene Name
protein artemis-like
There are no transcripts associated with this gene.

Gene
OLAH
Gene Name
oleoyl-ACP hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039702.2 Intron NP_001034791.1
NM_018324.2 Intron NP_060794.1
XM_017016376.1 Intron XP_016871865.1

View Full Product Details