Product Details

SNP ID
rs8679
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:226360853 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTCAAGAGCTCCCATGTTCAGT[A/G]TTCCTGGAGAAGGAAAGCTCTTTTT
Phenotype
MIM: 173870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARP1 PubMed Links

Gene Details

Gene
PARP1
Gene Name
poly(ADP-ribose) polymerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001618.3 3823 UTR 3 NP_001609.2

View Full Product Details