Product Details

SNP ID
rs3121819
Assay Type
Validated
NCBI dbSNP Submissions
49
Location
Chr.1:2020388 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCAAAGAGCCATCCCCCAGCATTC[G/A]GGATCTCAGAGACCGTGAAGATGTA
Phenotype
MIM: 137163
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GABRD PubMed Links

Gene Details

Gene
GABRD
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000815.4 Intron NP_000806.2
XM_011541194.2 Intron XP_011539496.1
XM_017000936.1 Intron XP_016856425.1
Gene
LOC105378589
Gene Name
uncharacterized LOC105378589
There are no transcripts associated with this gene.

View Full Product Details