Product Details

SNP ID
rs6062869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63255232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGGGTCTGGATGGGGCTGAA[C/T]CCCGTAAAGCCTGCGCTTCCGGAAG
Phenotype
MIM: 612873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ16779 PubMed Links

Gene Details

Gene
FLJ16779
Gene Name
uncharacterized LOC100192386
There are no transcripts associated with this gene.

Gene
LOC107985425
Gene Name
uncharacterized LOC107985425
There are no transcripts associated with this gene.

Gene
NKAIN4
Gene Name
Na+/K+ transporting ATPase interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152864.3 Intron NP_690603.3
XM_005260192.1 Intron XP_005260249.1
XM_005260194.3 Intron XP_005260251.1
XM_011528527.1 Intron XP_011526829.1
XM_011528528.2 Intron XP_011526830.1
XM_011528529.2 Intron XP_011526831.1
XM_017027636.1 Intron XP_016883125.1
XM_017027637.1 Intron XP_016883126.1
XM_017027638.1 Intron XP_016883127.1
XM_017027639.1 Intron XP_016883128.1

View Full Product Details