Product Details

SNP ID
rs35279523
Assay Type
Functionally Tested
NCBI dbSNP Submissions
15
Location
Chr.1:1243155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCCTGCAGCTCACTGTGGTCT[G/A]CGGAGAGAGCCCTGGGGAGGGTGGT
Phenotype
MIM: 615291 MIM: 616593
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
B3GALT6 PubMed Links
Additional Information
For this assay, SNP(s) [rs114921901] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
B3GALT6
Gene Name
beta-1,3-galactosyltransferase 6
There are no transcripts associated with this gene.

Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014980.2 782 Intron NP_001014980.1
XM_011541435.2 782 Missense Mutation XP_011539737.1
XM_011541436.2 782 Intron XP_011539738.1
XM_011541437.1 782 Nonsense Mutation XP_011539739.1

View Full Product Details