Product Details

SNP ID
rs3808964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123667111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCTAAGGCTCTGGGAACAGCC[G/T]CGCGGGATCTCGGCGGGAGTGGGAG
Phenotype
MIM: 604847
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR26 PubMed Links

Gene Details

Gene
GPR26
Gene Name
G protein-coupled receptor 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153442.3 Intron NP_703143.1

View Full Product Details