Product Details

SNP ID
rs869564
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:216635388 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAAAGGAAAAAGCCCCCACGAAATC[C/T]TTTCACTGCGGGGTGCCTGTCCCCC
Phenotype
MIM: 146731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFBP2 PubMed Links

Gene Details

Gene
IGFBP2
Gene Name
insulin like growth factor binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000597.2 Intron NP_000588.2
NM_001313990.1 Intron NP_001300919.1
NM_001313992.1 Intron NP_001300921.1
NM_001313993.1 Intron NP_001300922.1

View Full Product Details