Product Details

SNP ID
rs9333280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:182928670 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGCAGTTTAGGAAAAGGGAAAC[A/G]GACTACCCCATGTTCCACTGCCAGT
Phenotype
MIM: 604891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCKAP1 PubMed Links

Gene Details

Gene
NCKAP1
Gene Name
NCK associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013436.4 Intron NP_038464.1
NM_205842.2 Intron NP_995314.1
XM_006712200.3 Intron XP_006712263.1
XM_006712201.3 Intron XP_006712264.1

View Full Product Details