Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000909.5 | 2764 | Intron | NP_000900.1 | ||
XM_005263031.3 | 2764 | Intron | XP_005263088.1 | ||
XM_011532010.2 | 2764 | Intron | XP_011530312.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317091.1 | 2764 | Intron | NP_001304020.1 | ||
NM_001317092.1 | 2764 | Intron | NP_001304021.1 | ||
NM_006174.3 | 2764 | Intron | NP_006165.1 | ||
XM_005263038.3 | 2764 | UTR 5 | XP_005263095.1 | ||
XM_011532015.2 | 2764 | UTR 5 | XP_011530317.1 | ||
XM_011532017.2 | 2764 | UTR 5 | XP_011530319.1 | ||
XM_017008255.1 | 2764 | Intron | XP_016863744.1 | ||
XM_017008256.1 | 2764 | UTR 5 | XP_016863745.1 |