Product Details
- SNP ID
-
hCV310975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:58513116 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGGCCTACTTAAAATTGTAGCT[C/T]CCCCTCCTCCCTACTGACCCCCAGC
- Phenotype
-
MIM: 601641
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACOX2
PubMed Links
Gene Details
- Gene
- ACOX2
- Gene Name
- acyl-CoA oxidase 2
There are no transcripts associated with this gene.
View Full Product Details