Product Details

SNP ID
rs11769414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123925670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTGCATGAACCTGGCAGATGG[C/T]GGTTTTAGTGAGCCGAGATCATGCC
Phenotype
MIM: 600930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPAM1 PubMed Links

Gene Details

Gene
SPAM1
Gene Name
sperm adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174044.1 Intron NP_001167515.1
NM_001174045.1 Intron NP_001167516.1
NM_001174046.1 Intron NP_001167517.1
NM_003117.4 Intron NP_003108.2
NM_153189.2 Intron NP_694859.1

View Full Product Details