Product Details

SNP ID
rs10893304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124922277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACCAGTCCCTGGTGCCAAAAC[C/G]GTTGGGGACTGCTGCTATAAAGGGA
Phenotype
MIM: 611642 MIM: 611641
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HEPACAM PubMed Links

Gene Details

Gene
HEPACAM
Gene Name
hepatic and glial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152722.4 Intron NP_689935.2
XM_005271449.2 Intron XP_005271506.1
XM_017017361.1 Intron XP_016872850.1
Gene
HEPN1
Gene Name
hepatocellular carcinoma, down-regulated 1
There are no transcripts associated with this gene.

View Full Product Details