Product Details
- SNP ID
-
rs9881165
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:167490224 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CATATGACTCTCAAATAATGGTTGC[C/T]TGGGACCTGTTGAACCTTTTAAATA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
WDR49
PubMed Links
Gene Details
- Gene
- WDR49
- Gene Name
- WD repeat domain 49
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178824.3 |
|
Intron |
|
|
NP_849146.1 |
View Full Product Details