Product Details
- SNP ID
-
rs11707321
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:44980604 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CCTGCTGCAGTGAAAGTCATTTCTT[C/T]CATTTCCTCAGTAGGGATCCAGAGA
- Phenotype
-
MIM: 606488
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EXOSC7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28427135] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EXOSC7
- Gene Name
- exosome component 7
- Gene
- ZDHHC3
- Gene Name
- zinc finger DHHC-type containing 3
There are no transcripts associated with this gene.
View Full Product Details