Product Details

SNP ID
rs35959188
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:43100212 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GAGCCGGGAGAGACTGGAGCTTTGC[C/T]AATGGGGGCCTGGGCTGAGCAGGGC
Phenotype
MIM: 615247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POMK PubMed Links

Gene Details

Gene
POMK
Gene Name
protein-O-mannose kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277971.1 Intron NP_001264900.1
NM_032237.4 Intron NP_115613.1

View Full Product Details