Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001323573.1 | 3399 | Intron | NP_001310502.1 | ||
NM_001323574.1 | 3399 | Intron | NP_001310503.1 | ||
NM_001323575.1 | 3399 | Intron | NP_001310504.1 | ||
NM_032785.3 | 3399 | Intron | NP_116174.3 | ||
XM_011542308.2 | 3399 | Intron | XP_011540610.1 | ||
XM_011542310.2 | 3399 | Intron | XP_011540612.1 | ||
XM_017002595.1 | 3399 | UTR 3 | XP_016858084.1 | ||
XM_017002596.1 | 3399 | Intron | XP_016858085.1 | ||
XM_017002597.1 | 3399 | Intron | XP_016858086.1 | ||
XM_017002598.1 | 3399 | Intron | XP_016858087.1 |