Product Details

SNP ID
rs11556913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124451303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGGAGTCAAGTGGTCAGCCGAG[C/T]TGGAGGCTGATTTGCCCCGATCATG
Phenotype
MIM: 611244
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRMT12 PubMed Links

Gene Details

Gene
TRMT12
Gene Name
tRNA methyltransferase 12 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017956.3 497 Silent Mutation CTG,TTG L126L NP_060426.2

View Full Product Details