Product Details

SNP ID
rs229070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:26918618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTGAGTGTATAATGCAAAAATA[C/G]CATCCAGAGATTATATATTTCTTAA
Phenotype
MIM: 605007
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS5 PubMed Links

Gene Details

Gene
ADAMTS5
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007038.4 8957 UTR 3 NP_008969.2
XM_011529448.2 8957 UTR 3 XP_011527750.1

View Full Product Details