Product Details
- SNP ID
-
rs161826
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
62
- Location
-
Chr.1:7920278 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGATTTCAAAAAAACTAGCTCAA[A/G]CCTGTTAAGTGGTATCTAGAAAATG
- Phenotype
-
MIM: 602250
MIM: 604097
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TNFRSF9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201058455] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TNFRSF9
- Gene Name
- TNF receptor superfamily member 9
- Gene
- UTS2
- Gene Name
- urotensin 2
There are no transcripts associated with this gene.
View Full Product Details