Product Details

SNP ID
rs744115
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:11671120 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GGTTAACCCTTTGGTGTCCACAGCC[C/G]CAGAGACCTGAGCATGAGCAAGTCC
Phenotype
MIM: 609111 MIM: 605647 MIM: 604094
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXO44 PubMed Links

Gene Details

Gene
FBXO44
Gene Name
F-box protein 44
There are no transcripts associated with this gene.

Gene
FBXO6
Gene Name
F-box protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018438.5 Intron NP_060908.1
XM_005263448.4 Intron XP_005263505.1
XM_005263449.4 Intron XP_005263506.1
XM_005263451.4 Intron XP_005263508.1
XM_006710571.3 Intron XP_006710634.1
XM_011541232.2 Intron XP_011539534.1
XM_017001007.1 Intron XP_016856496.1
Gene
MAD2L2
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
There are no transcripts associated with this gene.

View Full Product Details