Product Details

SNP ID
rs619532
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:241629344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACTAATACACAATGCTGCTGCT[A/T]TAAGGACACATGCAATTAAAACAGT
Phenotype
MIM: 118825 MIM: 606695
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHML PubMed Links

Gene Details

Gene
CHML
Gene Name
CHM like, Rab escort protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001821.3 6587 UTR 3 NP_001812.2
Gene
OPN3
Gene Name
opsin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014322.2 6587 Intron NP_055137.2

View Full Product Details