Product Details

SNP ID
rs4643041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108507480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCAAGATAAAATCCACTCATG[G/T]TGACTATCATAGTATTGAGATCCAG
Phenotype
MIM: 612878
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EXPH5 PubMed Links

Gene Details

Gene
EXPH5
Gene Name
exophilin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144763.1 8138 UTR 3 NP_001138235.1
NM_001144764.1 8138 UTR 3 NP_001138236.1
NM_001144765.1 8138 UTR 3 NP_001138237.1
NM_001308019.1 8138 UTR 3 NP_001294948.1
NM_015065.2 8138 UTR 3 NP_055880.2
XM_011542696.2 8138 UTR 3 XP_011540998.1
XM_011542698.2 8138 UTR 3 XP_011541000.1
XM_011542700.2 8138 UTR 3 XP_011541002.1
XM_017017397.1 8138 UTR 3 XP_016872886.1
XM_017017398.1 8138 UTR 3 XP_016872887.1
XM_017017399.1 8138 UTR 3 XP_016872888.1
XM_017017400.1 8138 UTR 3 XP_016872889.1
XM_017017401.1 8138 UTR 3 XP_016872890.1
XM_017017402.1 8138 UTR 3 XP_016872891.1
XM_017017403.1 8138 UTR 3 XP_016872892.1
XM_017017404.1 8138 UTR 3 XP_016872893.1
Gene
KDELC2
Gene Name
KDEL motif containing 2
There are no transcripts associated with this gene.

View Full Product Details