Product Details

SNP ID
rs694769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118897721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTCCAGGGGAATGGAGGGAGC[A/C]ATAACTTGAAGAAGGGGGGAAGGGT
Phenotype
MIM: 609004 MIM: 601613
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCL9L PubMed Links

Gene Details

Gene
BCL9L
Gene Name
B-cell CLL/lymphoma 9-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182557.2 6159 UTR 3 NP_872363.1
XM_005271511.2 6159 UTR 3 XP_005271568.1
XM_005271512.2 6159 Intron XP_005271569.1
XM_005271513.2 6159 Intron XP_005271570.1
XM_006718815.2 6159 UTR 3 XP_006718878.1
XM_011542756.2 6159 Intron XP_011541058.1
XM_017017572.1 6159 UTR 3 XP_016873061.1
XM_017017573.1 6159 Intron XP_016873062.1
XM_017017574.1 6159 Intron XP_016873063.1
Gene
CXCR5
Gene Name
C-X-C motif chemokine receptor 5
There are no transcripts associated with this gene.

Gene
LOC101929133
Gene Name
uncharacterized LOC101929133
There are no transcripts associated with this gene.

View Full Product Details