Product Details

SNP ID
rs426102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:39109974 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACAGGTGATATTAGCACCTACT[A/G]TGAAGTCACTAATTGGTCCCAAATT
Phenotype
MIM: 602731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FYB PubMed Links

Gene Details

Gene
FYB
Gene Name
FYN binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243093.1 Intron NP_001230022.1
NM_001465.4 Intron NP_001456.3
NM_199335.3 Intron NP_955367.1
XM_006714464.2 Intron XP_006714527.1
XM_006714465.2 Intron XP_006714528.1
XM_006714466.2 Intron XP_006714529.1
XM_011514008.2 Intron XP_011512310.1
XM_011514009.1 Intron XP_011512311.1
XM_011514010.1 Intron XP_011512312.1
XM_011514011.2 Intron XP_011512313.1
XM_011514012.2 Intron XP_011512314.1
XM_011514013.2 Intron XP_011512315.1
XM_017009316.1 Intron XP_016864805.1

View Full Product Details