Product Details

SNP ID
rs404122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:39107251 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCTCACTAATGTAGTCTTCTGAG[A/T]TAACAATTAAGCAGAGAGATTATTT
Phenotype
MIM: 602731
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FYB PubMed Links

Gene Details

Gene
FYB
Gene Name
FYN binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243093.1 2668 UTR 3 NP_001230022.1
NM_001465.4 2668 UTR 3 NP_001456.3
NM_199335.3 2668 UTR 3 NP_955367.1
XM_006714464.2 2668 UTR 3 XP_006714527.1
XM_006714465.2 2668 UTR 3 XP_006714528.1
XM_006714466.2 2668 UTR 3 XP_006714529.1
XM_011514008.2 2668 UTR 3 XP_011512310.1
XM_011514009.1 2668 UTR 3 XP_011512311.1
XM_011514010.1 2668 UTR 3 XP_011512312.1
XM_011514011.2 2668 UTR 3 XP_011512313.1
XM_011514012.2 2668 Intron XP_011512314.1
XM_011514013.2 2668 Intron XP_011512315.1
XM_017009316.1 2668 UTR 3 XP_016864805.1

View Full Product Details