Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164326.1 | Intron | NP_001157798.1 | |||
NM_001300961.1 | Intron | NP_001287890.1 | |||
NM_001300962.1 | Intron | NP_001287891.1 | |||
NM_001300966.1 | Intron | NP_001287895.1 | |||
NM_006696.3 | Intron | NP_006687.3 | |||
NM_139199.1 | Intron | NP_631938.1 | |||
XM_005271855.2 | Intron | XP_005271912.1 | |||
XM_005271856.2 | Intron | XP_005271913.1 | |||
XM_005271857.2 | Intron | XP_005271914.1 | |||
XM_005271859.3 | Intron | XP_005271916.1 | |||
XM_005271860.3 | Intron | XP_005271917.1 | |||
XM_005271861.3 | Intron | XP_005271918.1 | |||
XM_011543109.1 | Intron | XP_011541411.1 | |||
XM_011543110.1 | Intron | XP_011541412.1 | |||
XM_017008969.1 | Intron | XP_016864458.1 | |||
XM_017008970.1 | Intron | XP_016864459.1 | |||
XM_017008971.1 | Intron | XP_016864460.1 | |||
XM_017008972.1 | Intron | XP_016864461.1 | |||
XM_017008973.1 | Intron | XP_016864462.1 | |||
XM_017008974.1 | Intron | XP_016864463.1 | |||
XM_017008975.1 | Intron | XP_016864464.1 | |||
XM_017008976.1 | Intron | XP_016864465.1 |