Product Details

SNP ID
rs618819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9891715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCAGCATTGGCCACTCTTGCCC[G/T]GATACAGCATGTATTAATGAGTTAA
Phenotype
MIM: 612087
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC2A PubMed Links

Gene Details

Gene
CLEC2A
Gene Name
C-type lectin domain family 2 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130711.1 Intron NP_001124183.1
NM_207375.2 Intron NP_997258.1
XM_011520654.2 Intron XP_011518956.1
XM_011520656.2 Intron XP_011518958.1
XM_011520657.2 Intron XP_011518959.1
Gene
KLRF2
Gene Name
killer cell lectin like receptor F2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190765.1 Intron NP_001177694.1

View Full Product Details