Product Details

SNP ID
rs232355
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:26911116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTAGCACCTTCCTTTCAGATC[C/T]TCTGGTCCTATAGATGCCTGAAGGT
Phenotype
MIM: 610191
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AQP4-AS1 PubMed Links

Gene Details

Gene
AQP4-AS1
Gene Name
AQP4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
CHST9
Gene Name
carbohydrate sulfotransferase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256316.1 6897 Intron NP_001243245.1
NM_031422.5 6897 Intron NP_113610.2
XM_005258362.3 6897 UTR 3 XP_005258419.1
XM_006722555.3 6897 Intron XP_006722618.1
XM_017026033.1 6897 UTR 3 XP_016881522.1
XM_017026034.1 6897 UTR 3 XP_016881523.1

View Full Product Details